Zinkoxid, NanoTek®

Lieferant: Thermo Scientific
Achtung

Synonyme: Zink(II)oxid

44899.22 44899.36 44899.A9
44899.22EA 47.5 EUR
44899.22 44899.36 44899.A9
Zinkoxid, NanoTek®
Zinkoxid

CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 40-100nm APS Powder, S.A. 10-25m2/g Density: 5.6 Odour: Odourless Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300


NanoTek® zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systems

Formel: ZnO
Molecular Weight: 81,39 g/mol
Schmelzpunkt: 1975 °C
Dichte: 5,6
Lagertemperatur: Raumtemperatur
MDL: MFCD00011300
CAS-Nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR