Zinksulfid ≥99,99% (Metall-Basis)
Lieferant: Thermo Scientific
Synonyme:
Zink(II)sulfid, Schwefelzink
13239.18
13239.30
13239.A1
13239.18EA
111
EUR
13239.18
13239.30
13239.A1
Zinksulfid ≥99,99% (Metall-Basis)
Zinksulfid
Purity: 99.99%
CAS No.: 1314-98-3
Molecular Formula: SZn
Formula Weight: 97.43
Physical Form: 2-20mm Lump
Density: 4.10
Appearance: White
Odour: Odourless
Melting Point: 1700°
Merck Reference: 14,10160
MDL No.: MFCD00011301
Formel:
SZn Molecular Weight: 97,46 g/mol Schmelzpunkt: 1700 °C Dichte: 4,10 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011301 CAS-Nummer: 1314-98-3 EINECS: 215-251-3 Merck Index: 14,10160 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...