Cadmium(II)selenid ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Cadmium (II) selenide
45456.18 45456.09
45456.18EA 536 EUR
45456.18 45456.09
Cadmium(II)selenid ≥99,999% (Metall-Basis)
Cadmium(II)selenid
Purity: 99.999% CAS No.: 1306-24-7 Molecular Formula: CdSe Formula Weight: 191.36 Physical Form: 3-12mm Pieces Density: 5.81 Appearance: Brown-red to black Odour: Odourless Melting Point: 1350° Merck Reference: 14,1626 UN No.: 3077 MDL No.: MFCD00010917
Formel: CdSe
Molecular Weight: 191,37 g/mol
Schmelzpunkt: >1350 °C
Dichte: 5,81
Lagertemperatur: Raumtemperatur
MDL: MFCD00010917
CAS-Nummer: 1306-24-7
EINECS: 215-148-3
UN: 3283
ADR: 6.1,III
Merck Index: 14,1626