Zinkselenid
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Zinc (II) selenide
A19947.09
A19947.18
A19947.30
A19947.09EA
27.4
EUR
A19947.09
A19947.18
A19947.30
Zinkselenid
Zinkselenid
CAS No.: 1315-09-9
Molecular Formula: ZnSe
Formula Weight: 144.33
Odour: Odourless
Melting Point:
Formel:
SeZn Molecular Weight: 144,35 g/mol Schmelzpunkt: >1100 °C Dichte: 5,42 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011296 CAS-Nummer: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1,III Merck Index: 14,10156 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...