Cadmium(II)sulfat, wasserfrei 99+% ACS
Lieferant: Thermo Scientific
Gefahr
11861.14 11861.22
11861.14EA 29.1 EUR
11861.14 11861.22
Cadmium(II)sulfat, wasserfrei 99+% ACS
Cadmium(II)sulfat
Purity: 99+% CAS No.: 10124-36-4 Molecular Formula: CdSO4 Formula Weight: 208.46 Physical Form: Powder Density: 4.691 Appearance: White Odour: Odourless Melting Point: 1000° Merck Reference: 14,1627 UN No.: 2570 MDL No.: MFCD00010923
Formel: CdO₄S
Molecular Weight: 208,47 g/mol
Schmelzpunkt: 1000 °C
Dichte: 4,691
Lagertemperatur: Raumtemperatur
MDL: MFCD00010923
CAS-Nummer: 10124-36-4
EINECS: 233-331-6
UN: 2570
ADR: 6.1,III
Merck Index: 14,1627