Yttrium(III)chlorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Fisher Scientific
Achtung

18682.18 18682.09 18682.30
18682.18EA 511 EUR
18682.18 18682.09 18682.30
Yttrium(III)chlorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Yttrium(III)chlorid

Purity: 99.9% CAS No.: 10361-92-9 Molecular Formula: Cl3Y Formula Weight: 195.26 Physical Form: -20 Mesh Density: 2.67 Appearance: White to pale yellow Odour: Odourless Melting Point: 721° Merck Reference: 14,10107 MDL No.: MFCD00011470

Formel: Cl₃Y
Molecular Weight: 195.26 g/mol
Siedepunkt: 1488
Schmelzpunkt: 721 °C
Dichte: 2.67
Lagertemperatur: Raumtemperatur
MDL: MFCD00011470
CAS-Nummer: 10361-92-9
EINECS: 233-801-0
Merck Index: 14,10107

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR