Zirkonium(IV)bromid ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Zirkon(IV)bromid

86139.09 86139.18
86139.09EA 104 EUR
86139.09 86139.18
Zirkonium(IV)bromid ≥99%
Zirkonium(IV)bromid

Purity: 99% CAS No.: 13777-25-8 Molecular Formula: ZrBr4 Formula Weight: 410.86 Physical Form: -40 Mesh Powder Density: 4.201 Melting Point: 450°/15 atm UN No.: 3260 MDL No.: MFCD00049637

Formel: Br₄Zr
Molecular Weight: 410,84 g/mol
Schmelzpunkt: 450 °C/15 atm
Dichte: 4,201
Lagertemperatur: Raumtemperatur
MDL: MFCD00049637
CAS-Nummer: 13777-25-8
EINECS: 237-417-4
UN: 3260
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR