Zirkonium(IV)fluorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Zirkontetrafluorid

86112.18 86112.30
86112.18EA 90.5 EUR
86112.18 86112.30
Zirkonium(IV)fluorid ≥98%
Zirkonium(IV)fluorid

Purity: 98% CAS No.: 7783-64-4 Molecular Formula: F4Zr Formula Weight: 167.21 Physical Form: Powder Density: 4.43 Refractive Index: 1.59 Sensitivity: Moisture Sensitive Appearance: White Melting Point: 600° subl. Solubility: Slightly soluble in water. Soluble in hydrofluoric acid Merck Reference: 14,10175 UN No.: 3260 MDL No.: MFCD00011307

Formel: F₄Zr
Molecular Weight: 167,22 g/mol
Schmelzpunkt: 600 °C (Subl.)
Dichte: 4,43
Lagertemperatur: Raumtemperatur
MDL: MFCD00011307
CAS-Nummer: 7783-64-4
EINECS: 232-018-1
UN: 3260
ADR: 8,II
Merck Index: 14,10175

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR