Titannitrid ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
Synonyme: Titanium nitride
41556.18 41556.30 41556.A1
41556.18EA 93.1 EUR
41556.18 41556.30 41556.A1
Titannitrid ≥99,5% (Metall-Basis)
Titannitrid
Purity: 99.5% CAS No.: 25583-20-4 Molecular Formula: TiN Formula Weight: 61.91 Physical Form: -325 Mesh Powder Density: 5.22 Appearance: Yellow Melting Point: 2950° MDL No.: MFCD00049596
Formel: NTi
Molecular Weight: 61,89 g/mol
Schmelzpunkt: 2950 °C
Dichte: 5,22
Lagertemperatur: Raumtemperatur
MDL: MFCD00049596
CAS-Nummer: 25583-20-4
EINECS: 247-117-5