Titan(III,IV)oxid ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
Synonyme:
Titanium oxide
39000.14
39000.22
39000.14EA
117
EUR
39000.14
39000.22
Titan(III,IV)oxid ≥99,9% (Metall-Basis)
Titanium(III,IV) oxide
Purity: 99.9%
CAS No.: 12065-65-5
Molecular Formula: Ti3O5
Physical Form: 3-10mm Pieces
Density: 4.20
Odour: Odourless
MDL No.: MFCD00210650
Used for optical coatings
Formel:
O₅Ti₃ Molecular Weight: 223,64 g/mol Schmelzpunkt: 1800 °C Dichte: 4,20 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210650 CAS-Nummer: 12065-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...