Zirkonium(IV)hydroxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

464171-100G 464171-500G
SIAL464171-100GEA 60.2 EUR
SIAL464171-100G SIAL464171-500G
Zirkonium(IV)hydroxid, Sigma-Aldrich®
Zirkonium(IV)hydroxid
Formel: H₄O₄Zr
CAS-Nummer: 14475-63-9
EINECS: 238-472-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR