Niob(V)chlorid ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Niobpentachlorid
unter Argon abgefüllt
11548.14
11548.22
11548.36
11548.14EA
108
EUR
11548.14
11548.22
11548.36
Niob(V)chlorid ≥99,9% (Metall-Basis)
Niob(V)chlorid
Purity: 99.9%
CAS No.: 10026-12-7
Molecular Formula: Cl5Nb
Formula Weight: 270.20
Physical Form: -100 Mesh Powder, Packaged under argon
Density: 2.74
Appearance: Yellow
Odour: Acidic
Melting Point: 208.3°
Boiling Point: 248.2°
Merck Reference: 14,6557
UN No.: 3260
MDL No.: MFCD00011127
Formel:
Cl₅Nb Molecular Weight: 270,17 g/mol Siedepunkt: 248,2 °C Schmelzpunkt: 208,3 °C Dichte: 2,74 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011127 CAS-Nummer: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8,III Merck Index: 14,6557 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...