Niob(V)chlorid ≥99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Niobpentachlorid
unter Argon abgefüllt
51108.09
51108.22
51108.36
51108.09EA
51.1
EUR
51108.09
51108.22
51108.36
Niob(V)chlorid ≥99% (Metall-Basis)
Niob(V)chlorid
Purity: 99%
CAS No.: 10026-12-7
Molecular Formula: NbCl5
Formula Weight: 270.20
Physical Form: 15mm & Down Lumps, Packaged under argon
Density: 2.74
Melting Point: 208.3°
Boiling Point: 248.2°
Solubility: Decomposes in water. Soluble in alcohol, HCl, and Carbon tetrachloride
Merck Reference: 14,6557
UN No.: 3260
MDL No.: MFCD00011127
In preparation of pure niobium, and as an intermediate
Formel:
Cl₅Nb Molecular Weight: 270,17 g/mol Siedepunkt: 248,2 °C Schmelzpunkt: 208,3 °C Dichte: 2,74 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011127 CAS-Nummer: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8,III Merck Index: 14,6557 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...