Vanadium(III)fluorid

Lieferant: CHEMPUR

FL7774.10 FL7774.25
CHMPFL7774.10EA 0 EUR
CHMPFL7774.10 CHMPFL7774.25
Vanadium(III)fluorid
Vanadium(III)fluorid
Formel: F₃V
Schmelzpunkt: 1406 °C
Dichte: 3,363 g/cm³ (20 °C)
MDL: MFCD00042553
CAS-Nummer: 10049-12-4
EINECS: 233-169-6
UN: 2923
ADR: 8,II
Merck Index: 12,10058

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR