Vanadium(IV)fluorid ≥95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Vanadium tetrafluoride

11543.06 11543.14
11543.06EA 55.1 EUR
11543.06 11543.14
Vanadium(IV)fluorid ≥95%
Vanadium(IV)fluorid

Purity: 95% CAS No.: 10049-16-8 Molecular Formula: F4V Formula Weight: 126.94 Physical Form: Powder Density: 2.975 Appearance: Yellow to brown Melting Point: 325° dec. Solubility: Soluble in water (blue Colour), acetone (deep green Colour), and glacial acetic acid (blue-green Colour) UN No.: 2923 MDL No.: MFCD00042554

Formel: F₄V
Molecular Weight: 126,94 g/mol
Schmelzpunkt: 325 °C (Zers.)
Dichte: 2,975
Lagertemperatur: Raumtemperatur
MDL: MFCD00042554
CAS-Nummer: 10049-16-8
EINECS: 233-171-7
UN: 2923
ADR: 8,II
Merck Index: 12,10057

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR