Vanadium(IV)fluorid
Lieferant: FLUOROCHEM
Synonyme:
Vanadium tetrafluoride
002871-10G
002871-25G
FLUO002871-10GEA
180
EUR
FLUO002871-10G
FLUO002871-25G
Vanadium(IV)fluorid
Vanadium(IV)fluorid
Formel:
F₄V Molecular Weight: 126,94 g/mol Schmelzpunkt: 325 °C Dichte: 2,97 g/cm³ (23 °C) |
MDL:
MFCD00042554 CAS-Nummer: 10049-16-8 EINECS: 233-171-7 UN: 2923 ADR: 8,II Merck Index: 12,10057 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...