Vanadium(III)oxid ≥95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Vanadiumtrioxid
12301.30
12301.A1
12301.A7
12301.30EA
93.2
EUR
12301.30
12301.A1
12301.A7
Vanadium(III)oxid ≥95%
Vanadium(III)oxid
Purity: 95%
CAS No.: 1314-34-7
Molecular Formula: O3V2
Formula Weight: 149.88
Physical Form: Powder
Density: 4.87
Appearance: Black
Odour: Odourless
Melting Point: 1970°
Solubility: Insoluble in water
Merck Reference: 14,9923
UN No.: 3285
MDL No.: MFCD00011455
Formel:
O₃V₂ Molecular Weight: 149,88 g/mol Schmelzpunkt: 1970 °C Dichte: 4,87 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011455 CAS-Nummer: 1314-34-7 EINECS: 215-230-9 UN: 3285 ADR: 6.1,II Merck Index: 14,9923 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...