Chrom(III)fluorid
Lieferant: FLUOROCHEM
Synonyme:
Chromium trifluoride
004566-5G
004566-25G
FLUO004566-5GEA
272
EUR
FLUO004566-5G
FLUO004566-25G
Chrom(III)fluorid
Chrom(III)fluorid
Formel:
CrF₃ Schmelzpunkt: 1000 °C Dichte: 3,8 g/cm³ (20 °C) |
MDL:
MFCD00016045 CAS-Nummer: 7788-97-8 EINECS: 232-137-9 UN: 1756 ADR: 8,II Merck Index: 12,02279 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...