Mangan(II)bromid, wasserfrei 99% H₂O <0.3%, Pulver -100 Mesh
Lieferant: Thermo Scientific
Achtung
Synonyme: Mangandibromid
32736.09 32736.18
32736.09EA 82.1 EUR
32736.09 32736.18
Mangan(II)bromid, wasserfrei 99% H₂O <0.3%, Pulver -100 Mesh
Mangan(II)bromid
It can be used in place of palladium in the Stille reaction, which couples two carbon atoms using an organotin compound.
Hygroscopic. Incompatible with oxidizing and reducing agents. Store at room temperature. Handle under dry protective gas.
Formel: Br₂Mn
Molecular Weight: 214,75 g/mol
Schmelzpunkt: 698 °C
Dichte: 4,38
Lagertemperatur: Raumtemperatur
MDL: MFCD00011113
CAS-Nummer: 13446-03-2
EINECS: 236-591-9
Merck Index: 14,5725