Mangan(II)chlorid, ultra trocken ≥99,99% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Manganese dichloride

Unter Argon in Ampullen verpackt

42844.04 42844.09 42844.18
42844.04EA 42.9 EUR
42844.04 42844.09 42844.18
Mangan(II)chlorid, ultra trocken ≥99,99% (Metall-Basis)
Mangan(II)chlorid

Purity: 99.99% CAS No.: 7773-01-5 Molecular Formula: Cl2Mn Formula Weight: 125.84 Physical Form: Powder, Ampouled under argon Density: 2.977 Appearance: Pink Odour: Odourless Melting Point: 650° Boiling Point: 1190° Merck Reference: 14,5728 UN No.: 3288 MDL No.: MFCD00011114

Formel: MnCl₂
Molecular Weight: 125,84 g/mol
Siedepunkt: 1190 °C
Schmelzpunkt: 650 °C
Dichte: 2,977
Lagertemperatur: Raumtemperatur
MDL: MFCD00011114
CAS-Nummer: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1,III
Merck Index: 14,5728

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR