Mangan(II)carbonat (≥44% Mn)
Lieferant: Thermo Scientific
Synonyme: Manganous carbonate
14324.36 14324.A3
14324.36EA 41.2 EUR
14324.36 14324.A3
Mangan(II)carbonat (≥44% Mn)
Mangan(II)carbonat
CAS No.: 598-62-9 Molecular Formula: CMnO3 Formula Weight: 114.95 Physical Form: -200 Mesh Powder Density: 3.125 Appearance: Light brown to violet Odour: Odourless Melting Point: 350° dec. Solubility: Insoluble in water, alcohol. Soluble in dilute inorganic acids Merck Reference: 14,5726 MDL No.: MFCD00011116
Pigment, varnish drier
Formel: CMnO₃
Molecular Weight: 114,95 g/mol
Schmelzpunkt: 350 °C (Zers.)
Dichte: 3,125
Lagertemperatur: Raumtemperatur
MDL: MFCD00011116
CAS-Nummer: 598-62-9
EINECS: 209-942-9
Merck Index: 14,5726