Mangan(II)carbonat ≥99,9% (Metall-Basis ausgeschlossen Na)
Lieferant: Thermo Scientific
Synonyme:
Manganous carbonate
14344.30
14344.A1
14344.30EA
58.2
EUR
14344.30
14344.A1
Mangan(II)carbonat ≥99,9% (Metall-Basis ausgeschlossen Na)
Mangan(II)carbonat
Purity: 99.9%
CAS No.: 598-62-9
Molecular Formula: MnCO3
Formula Weight: 114.95
Physical Form: Powder
Density: 3.125
Appearance: Light brown to violet
Odour: Odourless
Melting Point: 350° dec.
Merck Reference: 14,5726
MDL No.: MFCD00011116
Formel:
CMnO₃ Molecular Weight: 114,95 g/mol Schmelzpunkt: 350 °C (Zersetzung) Dichte: 3,125 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011116 CAS-Nummer: 598-62-9 EINECS: 209-942-9 Merck Index: 14,5726 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...