Mangan(II)carbonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Manganous carbonate

M-Clarity™ Qualitätsstufe = MQ200

377449-1KG 377449-250G
SIAL377449-1KGEA 193 EUR
SIAL377449-1KG SIAL377449-250G
Mangan(II)carbonat, Sigma-Aldrich®
Mangan(II)carbonat
Formel: CMnO₃
Dichte: 3,12 g/cm³ (20 °C)
MDL: MFCD00011116
CAS-Nummer: 598-62-9
EINECS: 209-942-9
Merck Index: 13,05749

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR