Mangan(III)fluorid ≥98% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

13069.09 13069.18
13069.09EA 76.6 EUR
13069.09 13069.18
Mangan(III)fluorid ≥98% (Metall-Basis)
Mangan(III)fluorid

Purity: 98% CAS No.: 7783-53-1 Molecular Formula: MnF3 Formula Weight: 111.93 Physical Form: Crystalline Density: 3.54 Melting Point: dec. Merck Reference: 14,5741 UN No.: 3288 MDL No.: MFCD00016223

Formel: F₃Mn
Molecular Weight: 111,93 g/mol
Schmelzpunkt: Zersetzung
Dichte: 3,54
Lagertemperatur: Raumtemperatur
MDL: MFCD00016223
CAS-Nummer: 7783-53-1
EINECS: 232-006-6
UN: 3087
ADR: 5.1,II
Merck Index: 14,5741

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR