Mangan(III)fluorid
Lieferant: FLUOROCHEM
003264-25G
003264-100G
003264-250G
FLUO003264-25GEA
0
EUR
FLUO003264-25G
FLUO003264-100G
FLUO003264-250G
Mangan(III)fluorid
Mangan(III)fluorid
Formel:
F₃Mn Dichte: 3,54 g/cm³ (20 °C) |
MDL:
MFCD00016223 CAS-Nummer: 7783-53-1 EINECS: 232-006-6 UN: 3288 ADR: 6.1,III Merck Index: 12,05784 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...