Mangan(II)iodid, ultra trocken ≥99,99% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Manganous iodide

Unter Argon in Ampullen verpackt

35710.04 35710.09
35710.04EA 165 EUR
35710.04 35710.09
Mangan(II)iodid, ultra trocken ≥99,99% (Metall-Basis)
Mangan(II)iodid

Purity: 99.99% CAS No.: 7790-33-2 Molecular Formula: I2Mn Formula Weight: 308.75 Physical Form: Powder/Flakes, Ampouled under argon Density: 5.0 Appearance: White to pink to red Odour: Odourless Melting Point: 638° Merck Reference: 14,5732 MDL No.: MFCD00016225

Formel: I₂Mn
Molecular Weight: 308,75 g/mol
Schmelzpunkt: 638 °C
Dichte: 5,0
Lagertemperatur: Raumtemperatur
MDL: MFCD00016225
CAS-Nummer: 7790-33-2
EINECS: 232-201-6
Merck Index: 14,5732

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR