Mangan(III)oxid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dimangantrioxid

87791.36 87791.A3
87791.36EA 191 EUR
87791.36 87791.A3
Mangan(III)oxid ≥98%
Mangan(III)oxid

Purity: 98% CAS No.: 1317-34-6 Molecular Formula: Mn2O3 Formula Weight: 157.86 Physical Form: -325 Mesh Powder Density: 4.50 Appearance: Black Odour: Odourless Melting Point: 1080° dec. Merck Reference: 14,5737 MDL No.: MFCD00016217

Formel: Mn₂O₃
Molecular Weight: 157,87 g/mol
Schmelzpunkt: 1080 °C (Zersetzung)
Dichte: 4,50
Lagertemperatur: Raumtemperatur
MDL: MFCD00016217
CAS-Nummer: 1317-34-6
EINECS: 215-264-4
Merck Index: 14,5737

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR