Iridium(IV)bromid ≥99,99% (Metall-Basis) Ir ≥ 37.1%, Premion®

Lieferant: Thermo Scientific
Achtung

39493.02 39493.04
39493.02EA 151 EUR
39493.02 39493.04
Iridium(IV)bromid ≥99,99% (Metall-Basis) Ir ≥ 37.1%, Premion®
Iridium(IV)bromid

Purity: 99.99% CAS No.: 7789-64-2 Molecular Formula: IrBr4 Formula Weight: 511.84 Physical Form: Crystalline Odour: Acrid MDL No.: MFCD00049467

Formel: Br₄Ir
Molecular Weight: 511,84 g/mol
Schmelzpunkt: Zersetzung
Lagertemperatur: Raumtemperatur
MDL: MFCD00049467
CAS-Nummer: 7789-64-2
EINECS: 232-180-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR