Cobalt(II)chlorid, wasserfrei ≥99,7% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Cobaltdichlorid
12303.18 12303.30
12303.18EA 101 EUR
12303.18 12303.30
Cobalt(II)chlorid, wasserfrei ≥99,7% (Metall-Basis)
Cobalt(II)chlorid
Purity: 99.7% CAS No.: 7646-79-9 Molecular Formula: Cl2Co Formula Weight: 129.84 Physical Form: Powder Density: 3.367 Appearance: Pale blue Odour: Odourless Melting Point: 735° Boiling Point: 1049° Solubility: Soluble in water, alcohol, acetone, ether, glycerol, and pyridine Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00010938
Formel: Cl₂Co
Molecular Weight: 129,84 g/mol
Siedepunkt: 1049 °C
Schmelzpunkt: 735 °C
Dichte: 3,367
Lagertemperatur: Raumtemperatur
MDL: MFCD00010938
CAS-Nummer: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 14,2437