Cobalt(II)chlorid, ultra trocken ≥99,998% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cobaltdichlorid
Unter Argon in Ampullen verpackt
13665.03
13665.06
13665.14
13665.03EA
97.9
EUR
13665.03
13665.06
13665.14
Cobalt(II)chlorid, ultra trocken ≥99,998% (Metall-Basis)
Cobalt(II)chlorid
Purity: 99.998%
CAS No.: 7646-79-9
Molecular Formula: Cl2Co
Formula Weight: 129.84
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.367
Appearance: Pale blue
Odour: Odourless
Melting Point: 735°
Boiling Point: 1049°
Merck Reference: 14,2437
UN No.: 3077
MDL No.: MFCD00010938
Formel:
Cl₂Co Molecular Weight: 129,84 g/mol Siedepunkt: 1049 °C Schmelzpunkt: 735 °C Dichte: 3,367 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010938 CAS-Nummer: 7646-79-9 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 14,2437 |