Nickel(II)chlorid, wasserfrei 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Nickeldichlorid
B22085.14
B22085.22
B22085.36
B22085.14EA
32.3
EUR
B22085.14
B22085.22
B22085.36
Nickel(II)chlorid, wasserfrei 98%
Nickel(II)chlorid
Purity: 98%
CAS No.: 7718-54-9
Molecular Formula: Cl2Ni
Formula Weight: 129.60
Odour: Odourless
Melting Point: 1001°
UN No.: 3288
MDL No.: MFCD00011142
Formel:
Cl₂Ni Molecular Weight: 129,6 g/mol Schmelzpunkt: 1001 °C Dichte: 3,55 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011142 CAS-Nummer: 7718-54-9 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 14,6505 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...