Platin(II)chlorid 99,9% (≥73% Pt) (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Platinum dichloride
11044.03
11044.06
11044.03EA
179
EUR
11044.03
11044.06
Platin(II)chlorid 99,9% (≥73% Pt) (Metall-Basis)
Platin(II)chlorid
Purity: 99.9%
CAS No.: 10025-65-7
Molecular Formula: Cl2Pt
Formula Weight: 266.00
Physical Form: Crystalline
Density: 5.87
Appearance: Green-brown
Melting Point: 581° dec.
Solubility: Insoluble in water, alcohol, ether. Soluble in HCl, NH4OH
Merck Reference: 14,7528
UN No.: 3261
MDL No.: MFCD00011181
Formel:
Cl₂Pt Molecular Weight: 265.99 g/mol Schmelzpunkt: 581 °C (Zers.) Dichte: 5.87 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011181 CAS-Nummer: 10025-65-7 EINECS: 233-034-1 UN: 3260 ADR: 8 III Merck Index: 14,7528 |