Cobalt(III)fluorid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cobalttrifluorid
11490.14
11490.22
11490.14EA
87.8
EUR
11490.14
11490.22
Cobalt(III)fluorid ≥99%
Cobalt(III)fluorid
Purity: 99%
CAS No.: 10026-18-3
Molecular Formula: CoF3
Formula Weight: 115.93
Physical Form: Powder
Density: 3.88
Merck Reference: 14,2430
UN No.: 3085
MDL No.: MFCD00010942
Formel:
CoF₃ Schmelzpunkt: 927 °C Dichte: 3,88 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010942 CAS-Nummer: 10026-18-3 EINECS: 233-062-4 UN: 3085 ADR: 5.1,III Merck Index: 14,2430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...