Nickel(II)fluorid, wasserfrei 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Nickeldifluorid
13067.09
13067.18
13067.09EA
101
EUR
13067.09
13067.18
Nickel(II)fluorid, wasserfrei 97%
Nickel(II)fluorid
Purity: 97%
CAS No.: 10028-18-9
Molecular Formula: NiF2
Formula Weight: 96.71
Physical Form: Crystalline
Density: 4.72
Appearance: Pale yellow
Odour: Odourless
Solubility: Slightly soluble in water
Merck Reference: 14,6508
UN No.: 3288
MDL No.: MFCD00016263
Formel:
F₂Ni Molecular Weight: 96.69 g/mol Schmelzpunkt: 1360…1380 °C Dichte: 4.72 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016263 CAS-Nummer: 10028-18-9 EINECS: 233-071-3 UN: 3288 ADR: 6.1 III Merck Index: 14,6508 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...