Platin(II)iodid ≥99,99% (Metall-Basis) Pt ≥ 43.0%, Premion®

Lieferant: Thermo Scientific
Achtung

Synonyme: Platindiiodid

12170.01 12170.03 12170.06
12170.01EA 64.6 EUR
12170.01 12170.03 12170.06
Platin(II)iodid ≥99,99% (Metall-Basis) Pt ≥ 43.0%, Premion®
Platin(II)iodid

Purity: 99.99% CAS No.: 7790-39-8 Molecular Formula: PtI2 Formula Weight: 448.90 Physical Form: Powder Odour: Odourless MDL No.: MFCD00011183

Formel: I₂Pt
Molecular Weight: 448,89 g/mol
Schmelzpunkt: 325 °C (Zersetzung)
Dichte: 6,403 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011183
CAS-Nummer: 7790-39-8
EINECS: 232-204-2
Merck Index: 12,07688

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR