Cobalt(II,III)oxid ≥99,7% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.7%
CAS No.: 1308-06-1
Molecular Formula: Co3O4
Formula Weight: 240.80
Physical Form: -400 Mesh Powder
Density: 6.11
Appearance: Black
Melting Point: 900° dec.
Solubility: Soluble in acids and alkalies
Merck Reference: 14,2429
UN No.: 3077
MDL No.: MFCD00010939
Enamels, semiconductors, grinding wheels
Formel:
Co₃O₄ Dichte: 6,11 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010939 CAS-Nummer: 1308-06-1 EINECS: 215-157-2 UN: 3077 ADR: 9,III Merck Index: 14,2429 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...