Eisen(II,III)oxid ≥97% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Trieisentetraoxid
12374.A1
12374.A7
12374.C4
12374.A1EA
64.4
EUR
12374.A1
12374.A7
12374.C4
Eisen(II,III)oxid ≥97% (Metall-Basis)
Eisen(II,III)oxid
Purity: 97%
CAS No.: 1317-61-9
Molecular Formula: Fe3O4
Formula Weight: 231.54
Physical Form: -325 Mesh Powder
Density: 5.2
Refractive Index: 3.0
Appearance: Black
Odour: Odourless
Melting Point: 1538°
Merck Reference: 14,4040
MDL No.: MFCD00011010
Formel:
Fe₃O₄ Molecular Weight: 231,54 g/mol Schmelzpunkt: 1538 °C Dichte: 5,2 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011010 CAS-Nummer: 1317-61-9 EINECS: 215-277-5 Merck Index: 14,4040 |