Eisen(II)sulfid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: Ferrous sulphide

17422.09 17422.18
17422.09EA 75.4 EUR
17422.09 17422.18
Eisen(II)sulfid ≥99% (Metall-Basis)
Eisen(II)sulfid

Purity: 99% CAS No.: 1317-37-9 Molecular Formula: FeS (Fe typically 0.9) Formula Weight: 87.91 Physical Form: -100 Mesh Powder Density: 4.74 Appearance: Brown to black Odour: Odourless Melting Point: 1195° Solubility: Soluble in acids with H2S evolution Merck Reference: 14,4058 MDL No.: MFCD00011013

Formel: FeS
Molecular Weight: 87,91 g/mol
Schmelzpunkt: 1195 °C
Dichte: 4,74
Lagertemperatur: Raumtemperatur
MDL: MFCD00011013
CAS-Nummer: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9,III
Merck Index: 14,4058

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR