Nickelsulfid ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Nickel subsulphide, Nickel disulphane
35661.18
35661.09
35661.18EA
434
EUR
35661.18
35661.09
Nickelsulfid ≥99,9% (Metall-Basis)
Nickelsulfid
Purity: 99.9%
CAS No.: 12035-72-2
Molecular Formula: Ni3S2
Formula Weight: 240.19
Physical Form: -200 Mesh Powder
Appearance: Black
Melting Point: 797°
UN No.: 3077
MDL No.: MFCD00016266
Formel:
Ni₃S₂ Molecular Weight: 240,21 g/mol Schmelzpunkt: 797 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00217044 CAS-Nummer: 12035-72-2 EINECS: 234-829-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...