Span® 20, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Sorbitan monododecanoate, Arlacel 20, Sorbitan monolaurate
MQ200
S6635-1L
S6635-250ML
SIALS6635-1LEA
221
EUR
SIALS6635-1L
SIALS6635-250ML
Span® 20, Sigma-Aldrich®
Span® 20
Formel:
C₁₈H₃₄O₆ Molecular Weight: 346.46 g/mol Dichte: 1.05 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005365 CAS-Nummer: 1338-39-2 EINECS: 215-663-3 Merck Index: 12,08872 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...