Dimidiumbromid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,8-Diamino-5-methyl-6-phenylphenanthridiniumbromid, 2,7-Diamino-10-methyl-9-phenylphenanthridiniumbromid
B24818.01
B24818.03
B24818.01EA
73.9
EUR
B24818.01
B24818.03
Dimidiumbromid 95%
Dimidiumbromid
Purity: 95%
CAS No.: 518-67-2
Molecular Formula: C20H18BrN3
Formula Weight: 380.31
Melting Point: 230° dec.
MDL No.: MFCD00011757
Formel:
C₂₀H₁₈BrN₃ Molecular Weight: 380,29 g/mol Schmelzpunkt: 230 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011757 CAS-Nummer: 518-67-2 EINECS: 208-256-7 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...