2,6-Dichlorpurin

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dichloro-9H-purine

010789-5G 010789-25G
FLUO010789-5GEA 0 EUR
FLUO010789-5G FLUO010789-25G
2,6-Dichlorpurin
2,6-Dichlorpurin
Formel: C₅H₂Cl₂N₄
Molecular Weight: 189 g/mol
Schmelzpunkt: 181
MDL: MFCD00077725
CAS-Nummer: 5451-40-1
EINECS: 226-681-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR