6-Chlorpurin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine

A11202.03 A11202.06 A11202.14
A11202.03EA 25.4 EUR
A11202.03 A11202.06 A11202.14
6-Chlorpurin ≥99%
6-Chlorpurin

Purity: 99% CAS No.: 87-42-3 Molecular Formula: C5H3ClN4 Formula Weight: 154.56 Melting Point: 300° dec. Beilstein Reference: 5774 Merck Reference: 14,2161 MDL No.: MFCD00075825

Formel: C₅H₃ClN₄
Molecular Weight: 154.56 g/mol
Siedepunkt: 185…192 °C (2 mmHg)
Schmelzpunkt: >300 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075825
CAS-Nummer: 87-42-3
EINECS: 201-745-6
Merck Index: 13,02180

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR