2-Amino-6-chlorpurin 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Chlor-7H-purin-2-ylamin

149290010 149290050
ACRO149290010EA 35.9 EUR
ACRO149290010 ACRO149290050
2-Amino-6-chlorpurin 99+%
2-Amino-6-chlorpurin
Formel: C₅H₄ClN₅
Molecular Weight: 169.57 g/mol
Schmelzpunkt: >300 °C
MDL: MFCD00075252
CAS-Nummer: 10310-21-1
EINECS: 233-686-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
HPLC ≥99.0 %
Titration with HClO4 ≥99.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR