Adenin, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Gefahr
|
Synonyme:
Purin-6-amin, 9H-Purin-6-amin, Vitamin B4, 6-Aminopurin
1152-10
1152-100
1152-25
1152-10EA
103
EUR
1152-10
1152-100
1152-25
Adenin, Millipore®
Adenin
Formel:
C₅H₅N₅ Molecular Weight: 135.13 g/mol Dichte: 0.99741 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041790 CAS-Nummer: 73-24-5 EINECS: 200-796-1 UN: 2811 ADR: 6.1 III Merck Index: 13,00152 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...