Hypoxanthin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1H-Purin-6-on, 6-Purinon, Purin-6(1H)-on, 6-Hydroxypurin

M-Clarity™ Qualitätsstufe = MQ200

H9377-1G H9377-100G H9377-25G H9377-5G
SIALH9377-1GEA 63.1 EUR
SIALH9377-1G SIALH9377-100G SIALH9377-25G SIALH9377-5G
Hypoxanthin, Sigma-Aldrich®
Hypoxanthin
Formel: C₅H₄N₄O
Molecular Weight: 136,11 g/mol
Schmelzpunkt: > 310 °C
MDL: MFCD00005725
CAS-Nummer: 68-94-0
EINECS: 200-697-3
Merck Index: 13,04895

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR