Purin 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
7H-Imidazo(4,5-d)pyrimidin
131670010
AC13167.1
AC13167.10
131670050
ACRO131670010EA
193
EUR
ACRO131670010
MAYBAC13167.1
MAYBAC13167.10
ACRO131670050
Purin 99%
Purin
Formel:
C₅H₄N₄ Molecular Weight: 120.11 g/mol Siedepunkt: 265 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079221 CAS-Nummer: 120-73-0 EINECS: 204-421-2 Merck Index: 12,08129 |
Spezifikation Testergebnisse
Appearance | White to light yellow to beige Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...