7-Methyladenin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Amino-7-methylpurin

M-Clarity™ Qualitätsstufe = MQ200

666548-100MG 666548-500MG
SIAL666548-100MGEA 161 EUR
SIAL666548-100MG SIAL666548-500MG
7-Methyladenin, Sigma-Aldrich®
7-Methyladenin
Formel: C₆H₇N₅
Molecular Weight: 149,16 g/mol
MDL: MFCD00127790
CAS-Nummer: 935-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR