1-Methylxanthin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methyl-1H,3H-purin-2,6-dion, 3,7-Dihydro-1-methyl-1H-purin-2,6-dion, 1-Methyl-3,7-dihydro-2H-purin-2,6-dion, 2,6-Dihydroxy-1-methylpurin, 1-Methyl-2,6(1H,3H)-purindion, 1-Methyl-3,7-dihydro-1H-purin-2,6-dion

69720-250MG 69720-1G
SIAL69720-250MGEA 212 EUR
SIAL69720-250MG SIAL69720-1G
1-Methylxanthin, Sigma-Aldrich®
1-Methylxanthin
Formel: C₆H₆N₄O₂
Molecular Weight: 166,14 g/mol
MDL: MFCD00005561
CAS-Nummer: 6136-37-4
EINECS: 228-108-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR