7-Methylxanthin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 7-Methyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dihydro-7-methyl-1H-purin-2,6-dion, 7-Methyl-1H,3H-purin-2,6-dion, 2,6-Dihydroxy-7-methylpurin, 7-Methyl-3,7-dihydro-2H-purin-2,6-dion, 7-Methyl-2,6(1H,3H)-purindion

H29190.MF H29190.MD
H29190.MFEA 83.6 EUR
H29190.MF H29190.MD
7-Methylxanthin ≥98%
7-Methylxanthin

Purity: 98% CAS No.: 552-62-5 Molecular Formula: C6H6N4O2 Formula Weight: 166.14 Melting Point: 300° MDL No.: MFCD00037979

Formel: C₆H₆N₄O₂
Molecular Weight: 166.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037979
CAS-Nummer: 552-62-5
EINECS: 209-019-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR